Skip to Content
Merck
All Photos(1)

Key Documents

483788

Sigma-Aldrich

N-(tert-Butoxycarbonyl)-D-prolinal

97%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€83.80

€83.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€83.80

About This Item

Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€83.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

optical activity

[α]23/D +83°, neat

reaction suitability

reaction type: solution phase peptide synthesis

refractive index

n20/D 1.461 (lit.)

bp

228 °C (lit.)

density

1.059 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

[H]C(=O)[C@H]1CCCN1C(=O)OC(C)(C)C

InChI

1S/C10H17NO3/c1-10(2,3)14-9(13)11-6-4-5-8(11)7-12/h7-8H,4-6H2,1-3H3/t8-/m1/s1

InChI key

YDBPZCVWPFMBDH-MRVPVSSYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Balboni et al.
European journal of medicinal chemistry, 35(11), 979-988 (2001-01-04)
A series of thirty 2-(3-pyridylaminomethyl)azetidine, pyrrolidine and piperidine analogues as nicotinic acetylcholine receptor (nAChR) ligands was explored. In general, pyrrolidinyl and many azetidinyl compounds were found to bind with enhanced affinity relative to the piperidines. In the three series, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service