Skip to Content
Merck
All Photos(1)

Key Documents

483761

Sigma-Aldrich

Boc-L-prolinal

97%

Synonym(s):

(S)-N-Boc-pyrrolidine-2-carboxaldehyde, N-(tert-Butoxycarbonyl)-L-prolinal

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€67.80

€67.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€67.80

About This Item

Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€67.80


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

optical activity

[α]20/D −101°, c = 0.66 in chloroform

reaction suitability

reaction type: Boc solid-phase peptide synthesis

refractive index

n20/D 1.462 (lit.)

bp

211 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

[H]C(=O)[C@@H]1CCCN1C(=O)OC(C)(C)C

InChI

1S/C10H17NO3/c1-10(2,3)14-9(13)11-6-4-5-8(11)7-12/h7-8H,4-6H2,1-3H3/t8-/m0/s1

InChI key

YDBPZCVWPFMBDH-QMMMGPOBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Key building block for norsecurinine-type alkaloids and isaindigotidione carboskeleton.[1][2]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

135.0 °F - closed cup

Flash Point(C)

57.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 45, 2985-2985 (2004)
Ramon Alibés et al.
Organic letters, 6(11), 1813-1816 (2004-05-21)
The syntheses of securinine and (-)-allonorsecurinine have been achieved starting from easily available alpha-amino acid derivatives and using as key steps a RCM and a Heck reaction for the formation of rings D and C, respectively. [reaction: see text]

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service