Skip to Content
Merck
All Photos(1)

Documents

47749

Sigma-Aldrich

Methyl 3-formylbenzoate

≥98.0% (GC)

Synonym(s):

Methyl benzaldehyde-3-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8O3
CAS Number:
Molecular Weight:
164.16
Beilstein:
2690668
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (GC)

form

solid

mp

48-52 °C

SMILES string

COC(=O)c1cccc(C=O)c1

InChI

1S/C9H8O3/c1-12-9(11)8-4-2-3-7(5-8)6-10/h2-6H,1H3

InChI key

UVSBCUAQEZINCQ-UHFFFAOYSA-N

Application

Methyl 3-formylbenzoate may be used in the preparation of the following bioactive compounds:
  • meso-Tetrakis(3-carboxyphenyl)porphyrin via condensation with pyrrole.
  • Methyl 3-[4-(1-methyl-3-phenylureido)phenylaminomethyl]benzoate, which shows moderate SENP1 protease inhibition activity.
  • An N-methyl-sulfonylhydrazone derivative for use as anti-diabetic agent with good plasma stability.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Discovery of 1-[4-(N-benzylamino) phenyl]-3-phenylurea derivatives as non-peptidic selective SUMO-sentrin specific protease (SENP) 1 inhibitors.
Uno M, et al.
Bioorganic & Medicinal Chemistry Letters, 22(16), 5169-5173 (2012)
Tetraphilin: a four-helix proton channel built on a tetraphenylporphyrin framework.
Akerfeldt KS, et al.
Journal of the American Chemical Society, 114(24), 9656-9657 (1992)
Synthesis, solubility, plasma stability, and pharmacological evaluation of novel sulfonylhydrazones designed as anti-diabetic agents.
Zapata-Sudo G, et al.
Drug design, development and therapy, 10, 2869-2869 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service