Skip to Content
Merck
All Photos(1)

Key Documents

47589

Sigma-Aldrich

Fmoc-Arg-OH

≥96.0%

Synonym(s):

Nα-Fmoc-L-arginine, Nα-(9-Fluorenylmethoxycarbonyl)-L-arginine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H24N4O4
CAS Number:
Molecular Weight:
396.44
Beilstein:
4828015
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥96.0%

form

solid

optical activity

[α]/D +9.0±2.0°, c = 1% in DMF

mp

145-150 °C (lit.)
145-160 °C (dec.)

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

NC(=N)NCCC[C@H](NC(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C21H24N4O4/c22-20(23)24-11-5-10-18(19(26)27)25-21(28)29-12-17-15-8-3-1-6-13(15)14-7-2-4-9-16(14)17/h1-4,6-9,17-18H,5,10-12H2,(H,25,28)(H,26,27)(H4,22,23,24)/t18-/m0/s1

InChI key

DVBUCBXGDWWXNY-SFHVURJKSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maria Koivisto et al.
Frontiers in pharmacology, 13, 871569-871569 (2022-05-21)
Contractility is one of the most crucial functions of the heart because it is directly related to the maintenance of blood perfusion throughout the body. Both increase and decrease in contractility may cause fatal consequences. Therefore, drug discovery would benefit
Ramon Cesar Botigelli et al.
Frontiers in cell and developmental biology, 10, 938709-938709 (2022-10-04)
Several opportunities for embryo development, stem cell maintenance, cell fate, and differentiation have emerged using induced pluripotent stem cells (iPSCs). However, the difficulty in comparing bovine iPSCs (biPSCs) with embryonic stem cells (ESCs) was a challenge for many years. Here
James R Kornacki et al.
ACS chemical biology, 10(1), 157-164 (2014-09-10)
Epigenetic regulation is directed, in part, by the correlated placement of histone post-translational modifications, but the mechanisms controlling correlated modifications are incompletely understood. Correlations arise from crosstalk among modifications and are frequently attributed to protein-protein interactions that recruit enzymes to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service