Merck
  • Acetyltransferase p300/CBP associated Factor (PCAF) regulates crosstalk-dependent acetylation of histone H3 by distal site recognition.
Acetyltransferase p300/CBP associated Factor (PCAF) regulates crosstalk-dependent acetylation of histone H3 by distal site recognition.
ACS chemical biology (2014-09-10)
James R Kornacki, Andreea D Stuparu, Milan Mrksich
ABSTRACT
Epigenetic regulation is directed, in part, by the correlated placement of histone post-translational modifications, but the mechanisms controlling correlated modifications are incompletely understood. Correlations arise from crosstalk among modifications and are frequently attributed to protein-protein interactions that recruit enzymes to existing histone modifications. Here we report the use of a peptide array to discover acetyltransferase-mediated crosstalks. We show that p300/CBP associated factor (PCAF)/GCN5 activity depends on the presence of a distal arginine residue of its histone H3 substrate. Modifications to H3 Arg8 decrease PCAF acetylation of H3 Lys14, and kinetic data indicate that arginine citrullination has the strongest effect in decreasing acetylation. Mutagenesis experiments demonstrate that PCAF specifically interprets H3 Arg8 modifications through interaction with residue Tyr640 on the surface of its catalytic domain, and this interaction regulates Lys14 acetylation by substrate discrimination. PCAF discriminates modified peptides as well as semisynthetic proteins and reconstituted nucleosomes bearing Arg8 modifications. Together, this work describes a method for systematically mapping crosstalks and illustrates its application to the discovery and elucidation of novel PCAF crosstalks.
Product Description
Supelco
Aphidicolin, analytical standard
Sigma-Aldrich
Imidazole, ReagentPlus®, 99%
Sigma-Aldrich
N,N-Dimethylformamide, anhydrous, 99.8%
Sigma-Aldrich
Imidazole, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Imidazole, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Imidazole, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Imidazole, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Imidazole, ≥99% (titration), crystalline
Imidazole, European Pharmacopoeia (EP) Reference Standard
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Imidazole, puriss. p.a., ≥99.5% (GC)
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Acetone, analytical standard
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Aphidicolin from Nigrospora sphaerica, ≥98% (HPLC), powder
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
N,N-Dimethylformamide, biotech. grade, ≥99.9%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material