Skip to Content
Merck
All Photos(2)

Key Documents

475807

Sigma-Aldrich

Glycerol 1,3-diglycerolate diacrylate

technical grade

Synonym(s):

Triglycerol diacrylate

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
€50.80

€50.80


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
€50.80

About This Item

Linear Formula:
[H2C=CHCO2CH2CH(OH)CH2OCH2]2CH(OH)
CAS Number:
Molecular Weight:
348.35
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€50.80


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

contains

1,000 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.446 (lit.)

density

1.237 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OC(COCC(O)COC(=O)C=C)COCC(O)COC(=O)C=C

InChI

1S/C15H24O9/c1-3-14(19)23-9-12(17)7-21-5-11(16)6-22-8-13(18)10-24-15(20)4-2/h3-4,11-13,16-18H,1-2,5-10H2

InChI key

PSSYEWWHQGPWGA-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan Yu et al.
International journal of biological macromolecules, 78, 32-38 (2015-04-04)
This paper introduces a novel chemical treatment for achieving sustained shrink-resist performance on natural keratin fibers. The new treatment involves the controlled reduction of keratin in the cuticle region of the fiber, and the application of a water soluble diacrylate
Gülçin Günal et al.
Artificial cells, nanomedicine, and biotechnology, 46(1), 178-184 (2017-03-23)
Monodisperse silica microspheres with bimodal pore-size distribution were proposed as a high performance sorbent for DNA isolation in batch fashion under equilibrium conditions. The proposed sorbent including both macroporous and mesoporous compartments was synthesized 5.1 μm in-size, by a "staged shape
Jian Yang et al.
Biomacromolecules, 15(11), 3942-3952 (2014-10-09)
Oxidative stress in tissue can contribute to chronic inflammation that impairs wound healing and the efficacy of cell-based therapies and medical devices. We describe the synthesis and characterization of a biodegradable, thermoresponsive gel with intrinsic antioxidant properties suitable for the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service