436895
Glycerol dimethacrylate, mixture of isomers
technical grade, 85%, contains 200 ppm monomethyl ether hydroquinone as inhibitor
Synonym(s):
Glycerol 1,3-dimethacrylate, Glycerol bis(meth)acrylate
Select a Size
About This Item
Recommended Products
grade
technical grade
Quality Level
Assay
85%
form
liquid
contains
200 ppm monomethyl ether hydroquinone as inhibitor
refractive index
n20/D 1.472 (lit.)
bp
120 °C/1 mmHg (lit.)
density
1.12 g/mL at 25 °C (lit.)
storage temp.
2-8°C
SMILES string
CC(=C)C(=O)OCC(O)COC(=O)C(C)=C.CC(=C)C(=O)OCC(CO)OC(=O)C(C)=C
InChI
1S/2C11H16O5/c1-7(2)10(13)15-5-9(12)6-16-11(14)8(3)4;1-7(2)10(13)15-6-9(5-12)16-11(14)8(3)4/h2*9,12H,1,3,5-6H2,2,4H3
InChI key
YDASFTNANXDGLQ-UHFFFAOYSA-N
Related Categories
General description
Application
- Used as a diluent in the development of new dental resinous systems. It is used to form highly crosslinked polymer networks, which are essential for providing composites with high stiffness, hardness, and durability.
- Used to prepare polymer matrix, along with other methacrylate monomers such as diurethanedimethacrylate (DUDMA) and urethane dimethacrylate (UDMA) for the development of 3D-printable antimicrobial composite materials for various applications, including dentistry.
- Used in the synthesis of highly cross-linked microbeads with thermoresponsive properties. The presence of GDMA contributes to the cross-linking of the polymeric structure. Thermoresponsive microbeads have potential applications in the biomedical field, such as drug delivery systems or tissue engineering.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
235.4 °F - closed cup
Flash Point(C)
113 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service