Skip to Content
Merck
All Photos(1)

Key Documents

46916

Sigma-Aldrich

9-Fluorenylmethyl carbamate

≥99.0% (HPLC)

Synonym(s):

Fmoc-amide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H13NO2
CAS Number:
Molecular Weight:
239.27
Beilstein:
4188192
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (HPLC)

form

solid

mp

201-204 °C

solubility

dioxane: 15 mg/mL, clear, colorless

application(s)

peptide synthesis

SMILES string

NC(=O)OCC1c2ccccc2-c3ccccc13

InChI

1S/C15H13NO2/c16-15(17)18-9-14-12-7-3-1-5-10(12)11-6-2-4-8-13(11)14/h1-8,14H,9H2,(H2,16,17)

InChI key

ZZOKVYOCRSMTSS-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Choi Yi Li et al.
Chembiochem : a European journal of chemical biology, 20(1), 46-50 (2018-09-19)
Urokinase-type plasminogen activator (uPA) and tissue-type plasminogen activator (tPA) are two serine proteases that contribute to initiating fibrinolysis by activating plasminogen. uPA is also an important tumour-associated protease due to its role in extracellular matrix remodelling. Overexpression of uPA has
Simon J de Veer et al.
The Biochemical journal, 469(2), 243-253 (2015-05-20)
Laskowski inhibitors regulate serine proteases by an intriguing mode of action that involves deceiving the protease into synthesizing a peptide bond. Studies exploring naturally occurring Laskowski inhibitors have uncovered several structural features that convey the inhibitor's resistance to hydrolysis and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service