All Photos(1)
About This Item
Linear Formula:
C6H5CH(OH)CO2CH2C6H5
CAS Number:
Molecular Weight:
242.27
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
99%
form
solid
optical activity
[α]25/D +55°, c = 1 in chloroform
mp
104-107 °C (lit.)
functional group
ester
hydroxyl
phenyl
SMILES string
O[C@H](C(=O)OCc1ccccc1)c2ccccc2
InChI
1S/C15H14O3/c16-14(13-9-5-2-6-10-13)15(17)18-11-12-7-3-1-4-8-12/h1-10,14,16H,11H2/t14-/m0/s1
InChI key
JFKWZVQEMSKSBU-AWEZNQCLSA-N
Application
Benzyl (S)-(+)-mandelate can be used as a reactant to prepare:
- α-Hydroxy-N-(2-phenylethyl)benzenepropanamide by reacting with benzeneethanamine.
- Benzyl-2-fluoro-2-phenylacetate by deoxyfluorination of alcohols.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Stereoselective Pseudomonas cepacia lipase mediated synthesis of α-hydroxyamides
Adamczyk M, et al.
Tetrahedron Asymmetry, 8(15), 2509-2512 (1997)
AlkylFluor: Deoxyfluorination of Alcohols.
Goldberg NW, et al.
Organic Letters, 18(23), 6102-6104 (2016)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service