If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf
Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
25 g | Check Cart for Availability | € 45,80 |
100 g | Check Cart for Availability | € 122,00 |
500 g | Check Cart for Availability | € 437,00 |
About This Item
grade
reagent grade
Quality Segment
product line
BioUltra
assay
≥99.0% (T)
reaction suitability
reagent type: catalyst
core: molybdenum
impurities
insoluble matter, passes filter test
pH
4.0-5.5 (25 °C, 0.05 M in H2O)
solubility
H2O: 0.05 M at 20 °C, clear, colorless
density
2.498 g/mL at 25 °C (lit.)
anion traces
chloride (Cl-): ≤10 mg/kg, nitrate (NO3-): ≤20 mg/kg, phosphate, silicate, arsenate (as PO43-): ≤10 mg/kg, sulfate (SO42-): ≤100 mg/kg
cation traces
Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Zn: ≤5 mg/kg
absorption
cut-off at 360 nm in H2O at 0.05 M
SMILES string
N.N.N.N.N.N.O.O.O.O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O=[Mo](=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O.O[Mo](O)(=O)=O
InChI
1S/7Mo.6H3N.10H2O.18O/h;;;;;;;6*1H3;10*1H2;;;;;;;;;;;;;;;;;;/q;;;;3*+2;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;;/p-6
InChI key
FIXLYHHVMHXSCP-UHFFFAOYSA-H
Application
- In situ preparation of molybdenum-dioxide-incorporated carbonized silk fiber and its application in supercapacitors.: This study discusses the in situ preparation of molybdenum dioxide incorporated into carbonized silk fiber, demonstrating its potential applications in supercapacitors due to enhanced electrical properties and stability (Ji et al., 2022).
- Constructing the Mo(2)C@MoO(x) Heterostructure for Improved SERS Application.: This paper presents the construction of a Mo(2)C@MoO(x) heterostructure, significantly improving surface-enhanced Raman scattering (SERS) applications due to its superior structural properties (Lai et al., 2022).
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| solubility H2O: 0.05 M at 20 °C, clear, colorless | solubility - | solubility - | solubility - |
| assay ≥99.0% (T) | assay 81.0-83.0% MoO3 basis, 99.98% trace metals basis | assay 81.0-83.0% MoO3 basis, 99.98% trace metals basis | assay 81.0-83.0% MoO3 basis |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| grade reagent grade | grade ACS reagent | grade ACS reagent | grade - |
| impurities insoluble matter, passes filter test | impurities ≤0.005% insolubles | impurities ≤0.005% insolubles | impurities - |
| cation traces Ca: ≤10 mg/kg, Co: ≤5 mg/kg, Cu: ≤5 mg/kg, K: ≤50 mg/kg, Mn: ≤5 mg/kg, Ni: ≤5 mg/kg, Zn: ≤5 mg/kg, Cd: ≤5 mg/kg, Fe: ≤5 mg/kg, Na: ≤50 mg/kg, Cr: ≤5 mg/kg, Pb: ≤5 mg/kg, Mg: ≤5 mg/kg | cation traces Mg and allied cations: ≤0.01%, heavy metals: ≤0.001% (as lead) | cation traces Mg and allied cations: ≤0.01%, heavy metals: ≤0.001% (as lead) | cation traces - |
| solubility H2O: 0.05 M at 20 °C, clear, colorless | solubility - | solubility - | solubility - |
| assay ≥99.0% (T) | assay 81.0-83.0% MoO3 basis, 99.98% trace metals basis | assay 81.0-83.0% MoO3 basis, 99.98% trace metals basis | assay 81.0-83.0% MoO3 basis |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| grade reagent grade | grade ACS reagent | grade ACS reagent | grade - |
| impurities insoluble matter, passes filter test | impurities ≤0.005% insolubles | impurities ≤0.005% insolubles | impurities - |
| cation traces Ca: ≤10 mg/kg, Co: ≤5 mg/kg, Cu: ≤5 mg/kg, K: ≤50 mg/kg, Mn: ≤5 mg/kg, Ni: ≤5 mg/kg, Zn: ≤5 mg/kg, Cd: ≤5 mg/kg, Fe: ≤5 mg/kg, Na: ≤50 mg/kg, Cr: ≤5 mg/kg, Pb: ≤5 mg/kg, Mg: ≤5 mg/kg | cation traces Mg and allied cations: ≤0.01%, heavy metals: ≤0.001% (as lead) | cation traces Mg and allied cations: ≤0.01%, heavy metals: ≤0.001% (as lead) | cation traces - |
Still not finding the right product?
Explore all of our products under Ammonium molybdate tetrahydrate
Storage Class
13 - Non Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-
