Skip to Content
Merck
All Photos(1)

Key Documents

452009

Sigma-Aldrich

2-Amino-4,5-diphenyl-3-furancarbonitrile

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12N2O
CAS Number:
Molecular Weight:
260.29
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

mp

201-205 °C (lit.)

functional group

nitrile
phenyl

SMILES string

Nc1oc(-c2ccccc2)c(-c3ccccc3)c1C#N

InChI

1S/C17H12N2O/c18-11-14-15(12-7-3-1-4-8-12)16(20-17(14)19)13-9-5-2-6-10-13/h1-10H,19H2

InChI key

BPFMLOQVCKVCAT-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure, Electrical Conductivity and Dielectric properties of bulk, 2-amino-(4, 5-diphenylfuran-3-carbonitrile).
Hendi AA.
Life Sciences, 8(3) (2011)
Fused Pyrimidines. Part II: Synthesis and Antimicrobial activity of Some Furo [3, 2-e] imidazo [1,2-c] pyrimidines and Furo[2,3-d]pyrimidines.
Bhuiyan MMH, et al.
Croatica Chemica Acta. Arhiv Za Kemiju, 78(4), 633-633 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service