Skip to Content
Merck
All Photos(1)

Key Documents

451215

Sigma-Aldrich

Praseodymium(III) chloride

anhydrous, beads, −10 mesh, 99.99% trace metals basis

Synonym(s):

Praseodymium chloride, Praseodymium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PrCl3
CAS Number:
Molecular Weight:
247.27
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

Assay

99.99% trace metals basis

form

beads

reaction suitability

reagent type: catalyst
core: praseodymium

impurities

≤150.0 ppm Trace Rare Earth Analysis

particle size

−10 mesh

mp

786 °C (lit.)

SMILES string

Cl[Pr](Cl)Cl

InChI

1S/3ClH.Pr/h3*1H;/q;;;+3/p-3

InChI key

LHBNLZDGIPPZLL-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service