Skip to Content
Merck
All Photos(1)

Key Documents

450723

Sigma-Aldrich

Bismuth(III) chloride

anhydrous, powder, 99.998% trace metals basis

Synonym(s):

Bismuth trichloride, Trichlorobismuth

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€105.00
25 G
€366.00

€105.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€105.00
25 G
€366.00

About This Item

Empirical Formula (Hill Notation):
BiCl3
CAS Number:
Molecular Weight:
315.34
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
powder

€105.00


Check Cart for Availability

Request a Bulk Order

grade

anhydrous

Quality Level

Assay

99.998% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: bismuth

impurities

≤15.0 ppm Trace Metal Analysis

bp

447 °C (lit.)

mp

230-232 °C (lit.)

SMILES string

Cl[Bi](Cl)Cl

InChI

1S/Bi.3ClH/h;3*1H/q+3;;;/p-3

InChI key

JHXKRIRFYBPWGE-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johanna Wagner et al.
Cell, 177(5), 1330-1345 (2019-04-16)
Breast cancer is a heterogeneous disease. Tumor cells and associated healthy cells form ecosystems that determine disease progression and response to therapy. To characterize features of breast cancer ecosystems and their associations with clinical data, we analyzed 144 human breast tumor
Yunqing Zhu et al.
ACS nano, 8(5), 5022-5031 (2014-04-09)
Homopolymers have been considered as a nonideal building block for creating well-defined nanostructures due to their fuzzy boundary between hydrophobic and hydrophilic moieties. However, this unique fuzzy boundary may provide some opportunities for fabricating functional nanomaterials. Presented in this paper

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service