Skip to Content
Merck
All Photos(1)

Key Documents

449385

Sigma-Aldrich

4-(Bromomethyl)benzophenone

96%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€174.00

€174.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€174.00

About This Item

Linear Formula:
BrCH2C6H4C(O)C6H5
CAS Number:
Molecular Weight:
275.14
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

€174.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

mp

109-112 °C (lit.)

functional group

bromo
ketone
phenyl

SMILES string

BrCc1ccc(cc1)C(=O)c2ccccc2

InChI

1S/C14H11BrO/c15-10-11-6-8-13(9-7-11)14(16)12-4-2-1-3-5-12/h1-9H,10H2

InChI key

RYULULVJWLRDQH-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph L Watson et al.
The Journal of cell biology, 220(2) (2021-01-09)
Protein micropatterning allows proteins to be precisely deposited onto a substrate of choice and is now routinely used in cell biology and in vitro reconstitution. However, drawbacks of current technology are that micropatterning efficiency can be variable between proteins and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service