Skip to Content
Merck
All Photos(1)

Key Documents

440639

Sigma-Aldrich

4,5-Dimethyl-2-furaldehyde

97%

Synonym(s):

4,5-Dimethylfurfural

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.529 (lit.)

bp

98-100 °C/12 mmHg (lit.)

density

1.016 g/mL at 25 °C (lit.)

functional group

aldehyde

SMILES string

Cc1cc(C=O)oc1C

InChI

1S/C7H8O2/c1-5-3-7(4-8)9-6(5)2/h3-4H,1-2H3

InChI key

JPTPEPVCVXGNJM-UHFFFAOYSA-N

General description

2,6-Dimethyl-4-pyrrone has been reported to undergo photochemical rearrangement to afford 4,5-dimethyl-2-furaldehyde. Mechanism of this photorearrangement has been described.[1][2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

150.8 °F - closed cup

Flash Point(C)

66 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photochemical Transformations of Small Ring Carbonyl Compounds. VIII. Photorearrangements in the Cyclopentenone Oxide Series1, 2.
Padwa A and Hartman R.
Journal of the American Chemical Society, 88(7), 1518-1524 (1966)
The photochemical rearrangement of 3, 4-diphenyl-4, 5-epoxy-2-cyclopentene-1-one.
Padwa A.
Tetrahedron Letters, 5(15), 813-813 (1964)
Cédric Peyrot et al.
Molecules (Basel, Switzerland), 25(9) (2020-05-10)
Faced with the ban of some organic UV filters such as octinoxate or avobenzone, especially in Hawaii, it became essential to offer new alternatives that are both renewable and safe for humans and the environment. In this context, a class
Xinyi Zhang et al.
Journal of agricultural and food chemistry, 67(12), 3502-3510 (2019-02-28)
This work outlines a rapid novel methodology for the direct quantitation of 12 volatile aldehyde compounds related to oxidative off-flavors in wine, by measuring the combined free and hydrogen-sulfite-bound forms of each aldehyde compound, consisting of four general aldehydes, four

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service