Skip to Content
Merck
All Photos(1)

Key Documents

526673

Sigma-Aldrich

5-Phenyl-2-furaldehyde

96%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€79.30

€79.30


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
1 G
€79.30

About This Item

Empirical Formula (Hill Notation):
C11H8O2
CAS Number:
Molecular Weight:
172.18
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€79.30


Estimated to ship on30 May 2025


Request a Bulk Order

Assay

96%

mp

29-33 °C (lit.)

functional group

aldehyde
phenyl

SMILES string

O=Cc1ccc(o1)-c2ccccc2

InChI

1S/C11H8O2/c12-8-10-6-7-11(13-10)9-4-2-1-3-5-9/h1-8H

InChI key

BMJHNNPEPBZULA-UHFFFAOYSA-N

General description

5-Phenyl-2-furaldehyde can be prepared from phenylfuran, via formylation.[1] It undergoes electrochemical reduction on a dropping Hg electrode to afford anion radicals in DMF.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Furan derivatives. XXXI.< x,/MJnsaturated ketones of the phenylfuran series.
Frimm R, et al.
Chem. Zeit., 27(1), 101-106 (1973)
Cathodic reduction of substituted 5-phenyl-2-furaldehydes in dimethylformamide.
Cernak J, et al.
Chemical Papers, 34(6), 788-792 (1980)
Renzo P Zanocco et al.
PloS one, 13(7), e0200006-e0200006 (2018-07-03)
In this study, we report the synthesis and the photochemical behavior of a series of new "click-on" fluorescent probes designed to detect singlet oxygen. They include a highly fluorescent chemical structure, an aryloxazole ring, linked to a furan moiety operating
Kyungsil Yoon et al.
Cells, 8(3) (2019-03-15)
Chicken ovalbumin upstream promoter-transcription factor I (COUP-TFI) is an orphan receptor and member of the nuclear receptor superfamily. Among a series of methylene substituted diindolylmethanes (C-DIMs) containing substituted phenyl and heteroaromatic groups, we identified 1,1-bis(3'-indolyl)-1-(4-pyridyl)-methane (DIM-C-Pyr-4) as an activator of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service