Skip to Content
Merck
All Photos(1)

Key Documents

439800

Sigma-Aldrich

Cadmium chloride

anhydrous, beads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Cadmium dichloride, Dichlorocadmium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CdCl2
Molecular Weight:
183.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

anhydrous

Quality Level

vapor density

6.3 (vs air)

vapor pressure

10 mmHg ( 656 °C)

Assay

99.999% trace metals basis

form

beads

reaction suitability

reagent type: catalyst
core: cadmium

particle size

−10 mesh

mp

568 °C (lit.)

solubility

H2O: soluble 457 g/L at 20 °C (OECD Test Guideline 105)

SMILES string

Cl[Cd]Cl

InChI

1S/Cd.2ClH/h;2*1H/q+2;;/p-2

InChI key

YKYOUMDCQGMQQO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Cadmium chloride (CdCl2) can be used for a variety of applications such as:
  • synthesis of cadmium selenide which can further be used in semiconductors(1)
  • synthesis of americium trichloride for potential usage in nuclear engineering(2)
  • a catalyst in the preparation of 5-substituted 1H-tetrazoles which can be utilized in pharmaceuticals, medicine, and agriculture(3)

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 1B - STOT RE 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yuxiong Huang et al.
PloS one, 15(9), e0239137-e0239137 (2020-10-01)
Sediment and soil contamination with toxic heavy metals, including cadmium (Cd2+) and lead (Pb2+), represents a major long-term remediation challenge. Resuspension of contaminated sediments into the water column, or the uptake of toxic metals from top soil, can lead to
Grace Choong et al.
Toxicology and applied pharmacology, 272(2), 423-430 (2013-07-23)
Cadmium disrupts the actin cytoskeleton in rat mesangial cells, and we have previously shown that this involves a complex interplay involving activation of kinase signaling, protein translocation, and disruption of focal adhesions. Here we investigate the role that glutathionylation of
Meesbah Jiwaji et al.
PloS one, 7(11), e50521-e50521 (2012-12-05)
In recent years much progress has been made in the development of tools for systems biology to study the levels of mRNA and protein, and their interactions within cells. However, few multiplexed methodologies are available to study cell signalling directly
M Auslander et al.
Marine biotechnology (New York, N.Y.), 10(3), 250-261 (2008-01-24)
Individuals of the fish Lithognathus mormyrus were exposed to a series of pollutants including: benzo[a]pyrene, pp-DDE, Aroclor 1254, perfluorooctanoic acid, tributyl-tin chloride, lindane, estradiol, 4-nonylphenol, methyl mercury chloride, and cadmium chloride. Five mixtures of the pollutants were injected. Each mixture
Veronique E Miron et al.
Nature neuroscience, 16(9), 1211-1218 (2013-07-23)
The lack of therapies for progressive multiple sclerosis highlights the need to understand the regenerative process of remyelination that can follow CNS demyelination. This involves an innate immune response consisting of microglia and macrophages, which can be polarized to distinct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service