Skip to Content
Merck
All Photos(1)

Key Documents

439487

Sigma-Aldrich

1,1,1,2,2,3,3,7,7,8,8,9,9,9-Tetradecafluoro-4,6-nonanedione

97%

Synonym(s):

5H,5H-Perdecafluoro-4,6-nonandione

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
€125.00

€125.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
€125.00

About This Item

Linear Formula:
(CF3CF2CF2CO)2CH2
CAS Number:
Molecular Weight:
408.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€125.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.323 (lit.)

bp

97 °C (lit.)

density

1.64 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(=O)CC(=O)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C9H2F14O2/c10-4(11,6(14,15)8(18,19)20)2(24)1-3(25)5(12,13)7(16,17)9(21,22)23/h1H2

InChI key

NICYPDXBQUERJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Coordination Chemistry, 38, 319-319 (1996)
Advanced Materials for Optics and Electronics, 1, 81-81 (1992)
In situ quadrupole mass spectroscopy studies of water and solvent coordination to copper (II) ?-diketonate precursors: implications for the chemical vapor deposition of copper.
Zheng B, et al.
Materials Chemistry and Physics, 41(3), 173-181 (1995)
Synthesis, characterization and crystal structure of a new thermally stable and volatile precursor [bis (1, 1, 1, 2, 2, 3, 3, 7, 7, 8, 8, 9, 9, 9-tetradecafluorononane-4, 6-dionato) 2-tetraglyme] barium (II) for MOCVD application.
Malandrino G, et al.
Journal of Materials Chemistry, 4(7), 1061-1066 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service