Skip to Content
Merck
All Photos(1)

Key Documents

426016

Sigma-Aldrich

4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione

99%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€31.10

€31.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€31.10

About This Item

Empirical Formula (Hill Notation):
C8H5F3O3
CAS Number:
Molecular Weight:
206.12
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€31.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.528 (lit.)

bp

203 °C (lit.)

mp

19-21 °C (lit.)

density

1.391 g/mL at 25 °C (lit.)

functional group

fluoro
ketone

storage temp.

2-8°C

SMILES string

FC(F)(F)C(=O)CC(=O)c1ccco1

InChI

1S/C8H5F3O3/c9-8(10,11)7(13)4-5(12)6-2-1-3-14-6/h1-3H,4H2

InChI key

OWLPCALGCHDBCN-UHFFFAOYSA-N

General description

4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione (furoyltrifluoroacetone, FTFA) is a β-diketone. Its cytotoxic activity against human cultured tumor and normal cells has been evaluated.[1] Reports suggest that 4,4,4-trifluoro-1-(2-furyl)-1,3-butanedione partially inhibits the oxidation of ferrocyanide in ETP (electron transport particles) isolated from beef heart mitochondria.[2] Its reaction with N,N,N′,N′-tetramethylalkyl diamines to form ionic adducts has been investigated.[3] The conformational analysis of the enol and keto form of FTFA has been reported.[4]

Application

4,4,4-Trifluoro-1-(2-furyl)-1,3-butanedione (tfa) may be used in the following studies:
  • As capping ligand in the synthesis of [Eu(tfa)3]2bpm complexes (bpm=2,2′-bipyrimidine).[5]
  • As reagent in the multistep synthesis of [13CD2]benzylamine.[6]
  • As reagent in the synthesis of 3-trifluoromethyl-2-arylcarbonylquinoxaline 1,4-di-N-oxide derivatives by reacting with corresponding benzofurazan oxides.[7]
  • In the efficient syntheses of perfluoroalkyl substituted azoles.[8]
  • Synthesis of 2-arylcarbonyl-3-trifluoromethylquinoxaline 1,4-di-N-oxide derivatives.[9]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kensuke Nakano et al.
Anticancer research, 24(2B), 711-717 (2004-05-27)
A variety of beta-diketones were evaluated for their cytotoxic profiles against oral human normal and tumor cells. Among 22 compounds (BD1-22) tested, the cytotoxicity of 3-formylchromone (BD17) (CC50=7.8 microg/mL) against human oral squamous cell carcinoma (HSC-2) cells was higher than
X-ray structure and temperature dependent luminescent properties of two bimetallic europium complexes.
Swavey S, et al.
Polyhedron, 27(3), 1061-1069 (2008)
An efficient entry to perfluoroalkyl substituted azoles starting from β-perfluoroalkyl-β-dicarbonyl compounds.
Bravo P, et al.
Tetrahedron, 50(29), 8827-8836 (1994)
Conformation, structure, intramolecular hydrogen bonding, and vibrational assignment of 4,4, 4-trifluoro-1-(2-furyl)-1,3-butanedione.
Tayyari SF, et al.
Journal of Molecular Structure, 8882(1), 153-167 (2008)
Acyclic tertiary diamines and 1, 4, 7,10-tetraazacyclododecane with fluorine-containing β-diketones: Leading to low melting ionic adducts.
Gupta OD, et al.
Journal of Fluorine Chemistry, 126(8), 1222-1229 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service