Skip to Content
Merck
All Photos(2)

Key Documents

43916

Sigma-Aldrich

Allyltris(3,3,4,4,5,5,6,6,6-nonafluorohexyl)stannane

~90%

Synonym(s):

Allyltris(1H,1H,2H,2H-perfluorohexyl)stannane, Tris(1H,1H,2H,2H-perfluorohexyl)allyltin

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€203.00

€203.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€203.00

About This Item

Empirical Formula (Hill Notation):
C21H17F27Sn
CAS Number:
Molecular Weight:
901.03
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

€203.00


Check Cart for Availability

Request a Bulk Order

Assay

~90%

form

liquid

density

1.670 g/mL at 25 °C

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)CC[Sn](CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F)(CCC(F)(F)C(F)(F)C(F)(F)C(F)(F)F)CC=C

InChI

1S/3C6H4F9.C3H5.Sn/c3*1-2-3(7,8)4(9,10)5(11,12)6(13,14)15;1-3-2;/h3*1-2H2;3H,1-2H2;

InChI key

TXGLPKAFRITSKI-UHFFFAOYSA-N

Legal Information

Product of Fluorous Technologies, Inc. Protected by U.S. patents 6,156,896; 5,859,247; and 5,777,126

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service