Skip to Content
Merck
All Photos(1)

Key Documents

434450

Sigma-Aldrich

Poly(1-vinylpyrrolidone-co-styrene)

38 % emulsion in H2O, <0.5 μm particle size

Synonym(s):

Styrene-N-Vinylpyrrolidone copolymer, Styrene-vinylpyrrolidinone copolymer, Styrene-vinylpyrrolidone copolymer, Vinylpyrrolidinone-styrene polymer

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€87.40

€87.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€87.40

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€87.40


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

composition

styrene, 64 wt. % (dry basis)

concentration

38 % emulsion in H2O

particle size

<0.5 μm

pH

2-5

viscosity

200-800 cP(25 °C, Brookfield, spindle #3) (20 rpm)(lit.)

density

1.04 g/mL at 25 °C (lit.)

SMILES string

N2(CCCC2=O)C=C.c1(ccccc1)C=C

InChI

1S/C8H8.C6H9NO/c1-2-8-6-4-3-5-7-8;1-2-7-5-3-4-6(7)8/h2-7H,1H2;2H,1,3-5H2

InChI key

ADNXYZUJPHVRPJ-UHFFFAOYSA-N

Features and Benefits

Stable, moderately viscous emulsion with high acid and salt tolerance. Forms strong, light-stable films with high water resistance.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service