Skip to Content
Merck
All Photos(4)

Documents

182729

Sigma-Aldrich

Poly(methyl vinyl ether) solution

50 wt. % in H2O

Synonym(s):

Methyl Vinyl Ether Polymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C3H6O)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

0.03 atm ( 20 °C)

concentration

50 wt. % in H2O

refractive index

n20/D 1.467

transition temp

Tg −31 °C

density

1.03 g/mL at 25 °C

SMILES string

COC=C

InChI

1S/C3H6O/c1-3-4-2/h3H,1H2,2H3

InChI key

XJRBAMWJDBPFIM-UHFFFAOYSA-N

Related Categories

Application

Poly(methyl vinyl ether) solution can be used asa precursor to synthesize thermosensitive microgels.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service