Skip to Content
Merck
All Photos(2)

Documents

429201

Sigma-Aldrich

Nylon 6/6

pellets

Synonym(s):

Poly(N,N′-hexamethyleneadipinediamide, Poly(hexamethylene adipamide)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C12H22N2O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

abrasion resistance

7 mg (Taber, CS-17 wheel, 1kg, 50% RH, 1,000 cycles)

form

pellets

autoignition temp.

752 °F

mol wt

~226.14

contains

≤23% proprietary impact modifier

composition

nylon, ≥76%

hardness

112 (Rockwell R, DAM, ASTM D 785)

mp

250-260 °C (lit.)

solubility

strong proton donor solvents, acetic acid and phenols: soluble

density

1.18 g/mL at 25 °C

SMILES string

NCCCCCCN.OC(=O)CCCCC(O)=O

InChI

1S/C12H24N2O2/c1-2-3-4-7-10-14-12(16)9-6-5-8-11(13)15/h2-10H2,1H3,(H2,13,15)(H,14,16)

InChI key

QDBQXOAICGSACD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Nylon 6/6 is a thermoplastic material with good mechanical properties like elastic modulus (~21 MPa) and peak stress (~6.5 MPa). It also has good thermal stability and flexibility that makes it useful in the packaging of food items.

Application

Nylon 6/6 can be used as nanofibers for the fabrication of support materials for osmosis membranes. It can also be used in the formation of the nanofibrous membranes as separators in lithium-ion batteries.

Features and Benefits

Impact resistant thermoplastic polyamide.
Stiff and strong over a wide range of temperatures, excellent toughness and chemical and abrasion resistance. Attacked by strong acids and strong bases at elevated temperatures. Oxidized by chlorine and hydrogen peroxide.

Other Notes

No lubricant added.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal and mechanical properties of electrospun PMMA, PVC, Nylon 6, and Nylon 6, 6
Carrizales C, et al.
Polymers For Advanced Technologies, 19(2), 124-130 (2008)
Tensile deformation of electrospun nylon-6, 6 nanofibers
Zussman E, et al.
Journal of Population Therapeutics and Clinical Pharmacology = Journal de la therapeutique Des Populations et de la Pharamcologie Clinique, 44(10), 1482-1489 (2006)
Sorption and transport of water vapor in nylon 6, 6 film
Lim L, et al.
Journal of Applied Polymer Science, 71(2), 197-206 (1999)
Hydrophilic nylon 6, 6 nanofibers supported thin film composite membranes for engineered osmosis
Huang, Liwei and McCutcheon, Jeffrey R
Journal of Membrane Science , 457(2), 162-169 (2014)
Evaluation of electrospun SiO2nylon 6, 6 nanofiber membranes as a thermally-stable separator for lithium-ion batteries
Yanilmaz M, et al.
Electrochimica Acta, 133(2), 501-508 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service