Skip to Content
Merck
All Photos(2)

Key Documents

429171

Sigma-Aldrich

Nylon 6/6

pellets

Synonym(s):

Poly(N,N′-hexamethyleneadipinediamide, Poly(hexamethylene adipamide)

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
€51.30
1 KG
€152.00

€51.30


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 G
€51.30
1 KG
€152.00

About This Item

Linear Formula:
(C12H22N2O2)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€51.30


Check Cart for Availability

Request a Bulk Order

abrasion resistance

4 mg (Taber, CS-17 wheel, 1kg, 50% RH, 1,000 cycles)

Quality Level

form

pellets

autoignition temp.

752 °F

mol wt

≥226.14

hardness

121 (Rockwell R, DAM, ASTM D 785)

relative viscosity

230-280(lit.)

mp

250-260 °C (lit.)

solubility

strong proton donor solvents, acetic acid and phenols: soluble

density

1.14 g/mL at 25 °C

SMILES string

NCCCCCCN.OC(=O)CCCCC(O)=O

InChI

1S/C12H24N2O2/c1-2-3-4-7-10-14-12(16)9-6-5-8-11(13)15/h2-10H2,1H3,(H2,13,15)(H,14,16)

InChI key

QDBQXOAICGSACD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Fibers for textiles, brushes and sutures cable sheathing and tubing and bearings, cams, gears and casings for tools and appliances.

Features and Benefits

Stiff and strong over a wide range of temperatures, excellent toughness and chemical and abrasion resistance. Attacked by strong acids and strong bases at elevated temperatures. Oxidized by chlorine and hydrogen peroxide.
High impact resistant thermoplastic polyamide with no impact modifier added. High viscosity for extrusion molding.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denisse Esther Mallaupoma Camarena et al.
Nanomaterials (Basel, Switzerland), 10(12) (2020-12-20)
The development of products for topical applications requires analyses of their skin effects before they are destined for the market. At present, the ban on animal use in several tests makes the search for in vitro models (such as artificial

Questions

1–4 of 4 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. What is the composition of the material? Which additives does it contain, if any?

    1 answer
    1. This material is a polyamide material with an estimated molecular weight of 20-50 kDa. The manufacturing process, including the presence of additives or binders, is considered proprietary.

      Helpful?

  4. 분자량 정보 확인 부탁드립니다. 스펙 상에 분자량이 ~226.14 로 되어 있는데 실제 평균 분자량 값이 맞는지 궁금합니다. 문헌상에는 해당 물질의 분자량이 보통 수만 g/mol 로 되어 있어서 확인 드립니다.

    1 answer
    1. The molecular formula posted to the product page - (C12H22N2O2)n - is for the monomer. The subscript 'n' indicates that this product is a polymer. The molecular weight would vary by batch and has not been determined. According to other sources the molecular weight can range from 20,000 to 50,000 Daltons.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service