Skip to Content
Merck
All Photos(1)

Key Documents

427128

Sigma-Aldrich

Gallium(III) chloride

anhydrous, beads, ≥99.999% trace metals basis

Synonym(s):

Gallium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
GaCl3
CAS Number:
Molecular Weight:
176.08
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
anhydrous
form:
beads
Pricing and availability is not currently available.

grade

anhydrous

vapor pressure

1 mmHg ( 48 °C)

Assay

≥99.999% trace metals basis

form

beads

reaction suitability

reagent type: catalyst
core: gallium

impurities

≤10.0 ppm Trace Metal Analysis

mp

78 °C (lit.)

density

2.47 g/mL at 25 °C (lit.)

SMILES string

Cl[Ga](Cl)Cl

InChI

1S/3ClH.Ga/h3*1H;/q;;;+3/p-3

InChI key

UPWPDUACHOATKO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Gallium(III) chloride is prepared by reacting Ga metal with Cl2 or HCl in N2 at 200oC.1

Application

Catalyst for a number of transformations such as the ring opening and recyclization of epoxides or the trimerization of alkynes.[1][2]

Features and Benefits

Lower-temperature solvothermal techniques using GaCl3 and NaN3 or Li3N result in nanocrystalline GaN.
Radical mediator and catalyst in a number of organic bond formation reactions.

Packaging

product is packed in vials

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chien-Hui Syu et al.
The Science of the total environment, 759, 143943-143943 (2020-12-20)
The emerging contaminants gallium (Ga) and indium (In) are extensively used in advanced industries and are considered as toxic to humans. Limited information is available on the dynamics of Ga and In in soil-upland crop systems. Therefore, this study aimed
Kee-Lung Chang et al.
Toxicology and applied pharmacology, 193(2), 209-217 (2003-12-03)
Gallium is commonly used in the semiconductor industry and medical field. Biologically, gallium is able to interrupt iron metabolism. Exposure to gallium has been shown to affect the human immune system. The purpose of this study was to investigate the
Katsukiyo Miura et al.
The Journal of organic chemistry, 72(3), 787-792 (2007-01-27)
The In(OAc)3-catalyzed reaction of bromo- and iodoalkanes with PhSiH3 in THF at 70 degrees C gave dehalogenated alkanes in good to high yields. In the presence of Et3B and air, the reduction proceeded smoothly at 30 degrees C. When 2,6-lutidine
E M Perchellet et al.
Anti-cancer drugs, 10(5), 477-488 (1999-09-07)
Gallium chloride (GaCl3), an antitumor agent with antagonistic action on iron, magnesium and calcium, was tested for its ability to alter the polymerization of purified tubulin (2.2 mg/ml) in a cell-free system in vitro. GaCl3 (250 microM) does not mimic
Muralidhar L Hegde et al.
Journal of molecular neuroscience : MN, 22(1-2), 19-31 (2004-01-27)
Previously, we evidenced a B --> Z helical change in Alzheimer's brain genomic DNA, leading to a hypothesis that Alzheimer's disease (AD) etiological factors such as aluminum (Al), amyloid beta (Abeta) peptide, and Tau might play a role in modulating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service