Skip to Content
Merck
All Photos(1)

Key Documents

427047

Sigma-Aldrich

Ethanol-2-13C

99 atom % 13C

Synonym(s):

13C Labeled ethanol, Ethyl alcohol-2-13C

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€1,030.00

€1,030.00


Check Cart for Availability

Request more information

Select a Size

Change View
1 G
€1,030.00

About This Item

Linear Formula:
13CH3CH2OH
CAS Number:
Molecular Weight:
47.06
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

€1,030.00


Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C

Quality Level

form

liquid

refractive index

n20/D 1.364 (lit.)

bp

78 °C (lit.)

mp

-114 °C (lit.)

density

0.801 g/mL at 25 °C

mass shift

M+1

SMILES string

[H]C([H])([13CH3])O

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3/i1+1

InChI key

LFQSCWFLJHTTHZ-OUBTZVSYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethanol-2-13C is an isotopic labeled precursor of ethanol.

Application

Ethanol-2-13C have been used as an internal standard to quantify the formed 2,4-dinitrophenylhydrazine using matrix-assisted laser desorption/ionization time-of-flight mass spectrometry (MALDI-TOFMS). It can be used for enzymatic oxidation and metabolic flux analyses.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Klaus Hollemeyer et al.
Rapid communications in mass spectrometry : RCM, 21(3), 336-342 (2007-01-09)
We describe a novel method for the determination of the concentration and labeling degree of ethanol originating from 1-13C-labeling experiments. This method is suitable for high-throughput metabolic flux analysis because of the possible parallel sample preparation and fast final analysis
Hong Liang et al.
Metabolic engineering, 65, 223-231 (2020-11-29)
Engineering microbes to utilize non-conventional substrates could create short and efficient pathways to convert substrate into product. In this study, we designed and constructed a two-step heterologous ethanol utilization pathway (EUP) in Escherichia coli by using acetaldehyde dehydrogenase (encoded by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service