Skip to Content
Merck
All Photos(2)

Documents

425257

Sigma-Aldrich

Tetramethylammonium nitrate

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4N(NO3)
CAS Number:
Molecular Weight:
136.15
Beilstein:
3917260
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

solid

mp

≥300 °C (lit.)

SMILES string

[O-][N+]([O-])=O.C[N+](C)(C)C

InChI

1S/C4H12N.NO3/c1-5(2,3)4;2-1(3)4/h1-4H3;/q+1;-1

InChI key

QGPVYHSXZFOSRL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Satendra Kumar et al.
Luminescence : the journal of biological and chemical luminescence, 33(3), 611-615 (2018-01-23)
Uranyl tris nitrato i.e. [UO2 (NO3 )3 ]- was formed by adding tetramethylammonium nitrate to uranyl nitrate in acetonitrile medium. The luminescence features of this complex in acetonitrile are very sensitive to water content, which could lead to the use
Sabrina Foglia et al.
Scientific reports, 7, 46513-46513 (2017-04-20)
Magnetic iron oxide nanoparticles (IONPs), for their intriguing properties, have attracted a great interest as they can be employed in many different biomedical applications. In this multidisciplinary study, we synthetized and characterized ultrafine 3 nm superparamagnetic water-dispersible nanoparticles. By a facile
Jacob O Fierer et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(13), E1176-E1181 (2014-03-19)
Individual proteins can now often be modified with atomic precision, but there are still major obstacles to connecting proteins into larger assemblies. To direct protein assembly, ideally, peptide tags would be used, providing the minimal perturbation to protein function. However
Nicholas D Lanz et al.
Biochemistry, 53(28), 4557-4572 (2014-06-06)
Lipoyl synthase (LS) catalyzes the final step in lipoyl cofactor biosynthesis: the insertion of two sulfur atoms at C6 and C8 of an (N(6)-octanoyl)-lysyl residue on a lipoyl carrier protein (LCP). LS is a member of the radical SAM superfamily
Lina Barreto et al.
Eukaryotic cell, 10(9), 1241-1250 (2011-07-05)
Potassium homeostasis is crucial for living cells. In the yeast Saccharomyces cerevisiae, the uptake of potassium is driven by the electrochemical gradient generated by the Pma1 H(+)-ATPase, and this process represents a major consumer of the gradient. We considered that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service