Skip to Content
Merck
All Photos(1)

Key Documents

421308

Sigma-Aldrich

MERPOL® HCS surfactant

Sign Into View Organizational & Contract Pricing

Select a Size

250 ML
€48.10

€48.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
250 ML
€48.10

About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

€48.10


Check Cart for Availability

Request a Bulk Order

form

liquid

Quality Level

refractive index

n20/D 1.428 (lit.)

bp

100 °C (lit.)

transition temp

cloud point 90 °C (1.7 wt. % aqueous solution)

density

1.03 g/mL at 25 °C (lit.)

HLB

15

Application

Nonionic, ethylene oxide condensate. General purpose detergent and wetting agent.

Legal Information

MERPOL is a registered trademark of Stepan Co.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service