Skip to Content
Merck
All Photos(1)

Key Documents

421170

Sigma-Aldrich

ZELEC® UN lubricant

technical grade

Synonym(s):

Nonionic Lubricant ZELEC UN

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€200.00

€200.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 L
€200.00

About This Item

MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

€200.00


Check Cart for Availability

Request a Bulk Order

grade

technical grade

Quality Level

form

liquid

refractive index

n20/D 1.445 (lit.)

bp

177 °C (lit.)

density

0.98 g/mL at 25 °C (lit.)

General description

ZELEC® UN lubricant is a colorless liquid, widely used as lubricant.

Legal Information

ZELEC is a registered trademark of Stepan Co.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

201.2 °F

Flash Point(C)

94 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service