Skip to Content
Merck
All Photos(5)

Key Documents

I19403

Sigma-Aldrich

Isophthaloyl chloride

≥99%

Synonym(s):

Isophthaloyl dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,3-(COCl)2
CAS Number:
Molecular Weight:
203.02
Beilstein:
638342
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

vapor pressure

0.03 mmHg ( 25 °C)

Quality Level

Assay

≥99%

expl. lim.

8.9 %

bp

276 °C (lit.)

mp

43-44 °C (lit.)

SMILES string

ClC(=O)c1cccc(c1)C(Cl)=O

InChI

1S/C8H4Cl2O2/c9-7(11)5-2-1-3-6(4-5)8(10)12/h1-4H

InChI key

FDQSRULYDNDXQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Isopthaloyl chloride acts as a crosslinking agent in various polymerization reactions.[1]

Application

Isopthaloyl chloride reacts with;
  • Polyethylene amine to produce PA-100 membrane[2]
  • Epiamine, a polyether amine, to produce PA-300 membrane[2]
  • 1,3-diamino-4-chlorobenzene to form polyamides[3]
  • Melamine forms polyamide aerogel[4]
  • bis-amine, produces an isophthalamide catenan[5]
  • Terephthaloyl chloride and bisphenol-type diazonaphthoquinone (DNQ) to polycondensate into a polyarylates containing DNQ structure[6]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

356.0 °F - closed cup

Flash Point(C)

180 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Constitutional isomerism in polyamides derived from isophthaloyl chloride and 1, 3-diamine-4-chlorobenzene.
Garcia JM, et al.
Journal of Polymer Science, 41(9), 1202-1215 (2003)
Rapid and facile synthesis of a low-cost monolithic polyamide aerogel via sol-gel technology
He S, et al.
Materials Letters, 144, 82-84 (2015)
Naylor T deV
Polymer Membranes: Materials, Structures and Separation Performance null
Sincero OA and Sincer GA
Physical-Chemical Treatment of Water and Wastewater null
Hyun Ju Oh et al.
Polymers, 12(7) (2020-07-28)
The colorimetric sensor is a facile, cost-effective, and non-power-operated green energy material for gas detection. In this study, the colorimetric sensing property of a meta-aramid/dye 3 nanofiber sensor for ammonia (NH3) gas detection was investigated. This colorimetric sensor was prepared

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service