Skip to Content
Merck
All Photos(1)

Key Documents

420670

Sigma-Aldrich

Ethyl 3-oxo-4-(triphenylphosphoranylidene)butyrate

97%

Synonym(s):

3-Oxo-4-(triphenylphosphoranylidene)butanoic acid ethyl ester, 4-(Triphenylphosphoranylidene)acetoacetic acid ethyl ester, [3-(Ethoxycarbonyl)-2-oxopropylidene]triphenylphosphorane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3P=CHCOCH2CO2C2H5
CAS Number:
Molecular Weight:
390.41
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

functional group

ester
ketone
phosphine

SMILES string

CCOC(=O)CC(=O)C=P(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C24H23O3P/c1-2-27-24(26)18-20(25)19-28(21-12-6-3-7-13-21,22-14-8-4-9-15-22)23-16-10-5-11-17-23/h3-17,19H,2,18H2,1H3

InChI key

QYXSFVCJCUXGJH-UHFFFAOYSA-N

Application

Can be coupled with glyoxals in a one-step route to 4-hydroxycyclopentanones. Also used to prepare 2H-pyran-2-ones from oxazolones.
Reactant for stereoselective synthesis of:
  • Polysubstituted cyclopentanones via double Michael addition reactions
  • Hydroindanes via asymmetric quadruple aminocatalytic three-component domino condensation and spirocyclization
  • Oxocyclohexenecarboxylates by cyclocondensation

Reactant for preparation of:
  • γ-(Dioxobutylidene)butenolides
  • Chain conjugated 2H-pyran-5-carboxylates
  • 2-Substituted pyridoacridines with antitumor activity by means of thermolysis

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 453-453 (1992)
Tetrahedron Letters, 34, 4837-4837 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service