Skip to Content
Merck
All Photos(1)

Key Documents

15038

Sigma-Aldrich

Boc-Arg(Pbf)-OH

≥98.0% (TLC)

Synonym(s):

Nα-Boc-Nω-(2,2,4,6,7-pentamethyl-dihydrobenzofuran-5-sulfonyl)-L-arginine, Nα-Boc-Nω-Pbf-L-arginine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€273.00

€273.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€273.00

About This Item

Empirical Formula (Hill Notation):
C24H38N4O7S
CAS Number:
Molecular Weight:
526.65
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

€273.00


Check Cart for Availability

Request a Bulk Order

Assay

≥98.0% (TLC)

form

powder

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

SMILES string

Cc1c(C)c(c(C)c2CC(C)(C)Oc12)S(=O)(=O)NC(=N)NCCC[C@H](NC(=O)OC(C)(C)C)C(O)=O

InChI

1S/C24H38N4O7S/c1-13-14(2)19(15(3)16-12-24(7,8)34-18(13)16)36(32,33)28-21(25)26-11-9-10-17(20(29)30)27-22(31)35-23(4,5)6/h17H,9-12H2,1-8H3,(H,27,31)(H,29,30)(H3,25,26,28)/t17-/m0/s1

InChI key

CVFXPOKENLGCID-KRWDZBQOSA-N

General description

may contain ~10% solvent

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hong Liang et al.
Small (Weinheim an der Bergstrasse, Germany), 16(10), e1906538-e1906538 (2020-02-06)
Codelivery of diagnostic probes and therapeutic molecules often suffers from intrinsic complexity and premature leakage from or degradation of the nanocarrier. Inspired by the "Y" shape of indocyanine green (ICG), the dye is integrated in an amphiphilic lipopeptide (RNF). The
Song Luo et al.
Colloids and surfaces. B, Biointerfaces, 148, 181-192 (2016-11-05)
A new drug delivery system with improved endocytosis efficiency based on arginine modified poly(ethylene glycol)-b-poly(ε-caprolactone) (PEG-PCL) diblock copolymers was developed successfully. The structures of amphiphilic copolymers were verified by proton Nuclear Magnetic Resonance (

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service