Skip to Content
Merck
All Photos(2)

Key Documents

405639

Sigma-Aldrich

2-Methyl-4′-(methylthio)-2-morpholinopropiophenone

98%

Synonym(s):

α-Methyl-4-(methylmercapto)-α-morpholinopropiophenone, 2-Methyl-1-[4-(methylthio)phenyl]-2-(4-morpholinyl)-1-propanone, 2-Methyl-2-(4-morpholinyl)-1-[4-(methylthio)phenyl]-1-propanone, 2-Methyl[4-(methylthio)phenyl]-2-morpholinopropan-1-one

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€103.00

€103.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 G
€103.00

About This Item

Empirical Formula (Hill Notation):
C15H21NO2S
CAS Number:
Molecular Weight:
279.40
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€103.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

mp

74-76 °C (lit.)

SMILES string

CSc1ccc(cc1)C(=O)C(C)(C)N2CCOCC2

InChI

1S/C15H21NO2S/c1-15(2,16-8-10-18-11-9-16)14(17)12-4-6-13(19-3)7-5-12/h4-7H,8-11H2,1-3H3

InChI key

LWRBVKNFOYUCNP-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>329.0 °F

Flash Point(C)

> 165 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mengfei Wang et al.
Nature communications, 8(1), 388-388 (2017-09-01)
Disclinations are topological singularities of molecular arrangement in liquid crystals, which typically occur when the average orientation of molecules makes a π rotation along a fictitious closed loop taken inside the liquid crystal. Depending on the sense of molecular rotation
Preston Sutton et al.
Polymers, 12(3) (2020-03-11)
Lithium metal anodes have been pursued for decades as a way to significantly increase the energy density of lithium-ion batteries. However, safety risks caused by flammable liquid electrolytes and short circuits due to lithium dendrite formation during cell cycling have
Po-Hsin Wang et al.
Nanomaterials (Basel, Switzerland), 8(4) (2018-04-13)
A photopolymerization method is used to prepare a mixture of polymer ionic liquid (PIL) and ionic liquid (IL). This mixture is used as a solid-state electrolyte in carbon nanoparticle (CNP)-based symmetric supercapacitors. The solid electrolyte is a binary mixture of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service