Skip to Content
Merck
All Photos(2)

Key Documents

406368

Sigma-Aldrich

2,4-Diethyl-9H-thioxanthen-9-one

98%

Synonym(s):

2,4-Diethylthioxanthone

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€97.10

€97.10


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€97.10

About This Item

Empirical Formula (Hill Notation):
C17H16OS
CAS Number:
Molecular Weight:
268.37
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€97.10


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

solid

mp

66-70 °C (lit.)

SMILES string

CCc1cc(CC)c2Sc3ccccc3C(=O)c2c1

InChI

1S/C17H16OS/c1-3-11-9-12(4-2)17-14(10-11)16(18)13-7-5-6-8-15(13)19-17/h5-10H,3-4H2,1-2H3

InChI key

BTJPUDCSZVCXFQ-UHFFFAOYSA-N

Application

Photoinitiator

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yinfeng He et al.
Journal of biomedical materials research. Part B, Applied biomaterials, 105(6), 1645-1657 (2016-05-15)
A new type of photocrosslinkable polycaprolactone (PCL) based ink that is suitable for three-dimensional (3D) inkjet printing has been developed. Photocrosslinkable Polycaprolactone dimethylacrylate (PCLDMA) was synthesized and mixed with poly(ethylene glycol) diacrylate (PEGDA) to prepare an ink with a suitable

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service