Skip to Content
Merck
All Photos(1)

Documents

402133

Sigma-Aldrich

6-Ethyl-3-formylchromone

98%

Synonym(s):

6-Ethyl-4-oxo-4H-1-benzopyran-3-carboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10O3
CAS Number:
Molecular Weight:
202.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

108-110 °C (lit.)

SMILES string

CCc1ccc2OC=C(C=O)C(=O)c2c1

InChI

1S/C12H10O3/c1-2-8-3-4-11-10(5-8)12(14)9(6-13)7-15-11/h3-7H,2H2,1H3

InChI key

XCKTXKYXKJJGBA-UHFFFAOYSA-N

General description

6-Ethyl-3-formylchromone (6-Ethyl-4-oxo-4H-1-benzopyran-3-carboxaldehyde) is a benzopyran derivative. It is a bicyclic heterocyclic molecule made up of a benzene ring fused to a heterocyclic pyran ring.

Application

6-Ethyl-3-formylchromone is the suitable reagent used in the synthesis of uridine-based library. It may be used in the synthesis of ethyl 3-acetyl-5-(5-ethyl-2-hydroxybenzoyl)-2-hydroxybenzoate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of polyketide-type phenols by domino 'Michael/retro-Michael/Aldol'reactions of 3-formylchromones with silyl enol ethers derived from ethyl 3, 5-dioxohexanoate.
Adeel M, et al.
Tetrahedron, 65(21), 4099-4105 (2009)
Howard C Hang et al.
Chemistry & biology, 11(3), 337-345 (2004-05-05)
The polypeptide N-acetyl-alpha-galactosaminyltransferases (ppGalNAcTs, also abbreviated ppGaNTases) initiate mucin-type O-linked glycosylation and therefore play pivotal roles in cell-cell communication and protection of tissues. In order to develop new tools for studying mucin-type O-linked glycosylation, we screened a 1338 member uridine-based

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service