Skip to Content
Merck
All Photos(1)

Key Documents

543624

Sigma-Aldrich

6-Fluorochromone-3-carboxaldehyde

97%

Synonym(s):

6-Fluoro-3-formylchromone

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€36.00

€36.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€36.00

About This Item

Empirical Formula (Hill Notation):
C10H5FO3
CAS Number:
Molecular Weight:
192.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€36.00


Check Cart for Availability

Request a Bulk Order

Assay

97%

mp

155-160 °C (lit.)

functional group

aldehyde
fluoro
ketone

SMILES string

Fc1ccc2OC=C(C=O)C(=O)c2c1

InChI

1S/C10H5FO3/c11-7-1-2-9-8(3-7)10(13)6(4-12)5-14-9/h1-5H

InChI key

VHRMOTNEBIKURN-UHFFFAOYSA-N

Gene Information

human ... PTPN1(5770)

Application

6-Fluorochromone-3-carboxaldehyde may be used in the synthesis of the following fluorine-containing chromone-tetrazoles:[1]
  • 3-((1-cyclohexyl-1H-tetrazol-5-yl)(phenylamino)methyl)-6-fluoro-4H-chromen-4-one
  • 3-((1-cyclohexyl-1H-tetrazol-5-yl)((3,4,5-trimethoxyphenyl)amino)methyl)-6-fluoro-4H-chrome-4-one
  • 3-((1-(tert-butyl)-1H-tetrazol-5-yl)(phenylamino)methyl)-6-fluoro-4H-chromen-4-one
  • 3-((1-(tert-butyl)-1H-tetrazol-5-yl)((4-chlorophenyl)amino)methyl)-6-fluoro-4H-chromen-4-one

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pedro A Cano et al.
Molecules (Basel, Switzerland), 20(7), 12436-12449 (2015-07-18)
In vitro studies to fourteen previously synthesized chromone-tetrazoles and four novel fluorine-containing analogs were conducted against pathogenic protozoan (Entamoeba histolytica), pathogenic bacteria (Pseudomonas aeruginosa, and Staphylococcus aureus), and human fungal pathogens (Sporothrix schenckii, Candida albicans, and Candida tropicalis), which have

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service