Skip to Content
Merck
All Photos(1)

Key Documents

393851

Sigma-Aldrich

2-Hydroxy-5-methyl-1,3-benzenedicarboxaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H2(CH3)(CHO)2
CAS Number:
Molecular Weight:
164.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

128-130 °C (lit.)

functional group

aldehyde

SMILES string

Cc1cc(C=O)c(O)c(C=O)c1

InChI

1S/C9H8O3/c1-6-2-7(4-10)9(12)8(3-6)5-11/h2-5,12H,1H3

InChI key

ZBOUXALQDLLARY-UHFFFAOYSA-N

General description

2-Hydroxy-5-methyl-1,3-benzenedicarboxaldehyde (2-hydroxy-5-methylisophthalaldehyde) is a dialdehyde derivative. It has been synthesized by heating p-cresol with hexamethylenetetramine. The structure has been confirmed by 1H and 13C NMR.[1] It is an important raw material for the synthesis of various binucleating schiff base ligand.[2]

Application

2-Hydroxy-5-methyl-1,3-benzenedicarboxaldehyde (2-hydroxy-5-methylisophthalaldehyde) is suitable reagent used in the synthesis of 2-(2′-vinyloxyethoxy)-5-methylisophthaldehyde[3] and chiral calixsalen macrocycles.[4]
2-Hydroxy-5-methyl-1,3-benzenedicarboxaldehyde may be used in the synthesis of the following:
  • 3-[(2,4-Dichlorophenyl)iminomethyl]-2-hydroxy-5-methylbenzaldehyde, a Schiff base.[5]
  • 2-Hydroxy-3-methoxymethyl-5-methylbenzaldehyde.[6]
  • Acyclic Schiff-base ligands.[7]
  • Macrobicyclic ligands (MSB).[8]
  • 4-Methyl-2,6-divinylphenol.[1]
  • 2-Hydroxy-3-dimethoxymethyl-5-methylbenzaldehyde.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

3-[(2, 4-Dichlorophenyl) iminomethyl]-2-hydroxy-5-methylbenzaldehyde.
Kilic I, et al
Acta Crystallographica Section E, Structure Reports Online, 65(6), 1347-1347 (2009)
Steric effect in the free radical polymerization of vinyl ethers containing electron-deficient olefin groups.
Lee JY and Jin MK
Polymer Bull., 44(3), 2777-2284 (2000)
New efficient ruthenium metathesis catalyst containing chromenyl ligand.
Hryniewicka A, et al
Journal of Organometallic Chemistry, 695(9), 1265-1270 (2010)
ESI MS, spectroscopic and semiempirical characterization of a macrobicyclic complex with Er (III) cation.
Przybylski P, et al.
Journal of Molecular Structure, 878(1), 95-103 (2008)
2-Hydroxy-3-methoxymethyl-5-methylbenzaldehyde.
Gunasekaran B, et al
Acta Crystallographica Section E, Structure Reports Online, 69(2), 317-317 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service