Skip to Content
Merck
All Photos(1)

Key Documents

924423

Sigma-Aldrich

2,4,6-Trihydroxybenzene-1,3,5-tricarbaldehyde

Synonym(s):

1,3,5-Triformyl-2,4,6-trihydroxybenzene, 1,3,5-Triformylphloroglucinol, 2,4,6-Triformylphloroglucinol, 2,4,6-Trihydroxy-1,3,5-benzenetricarboxaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€131.00

€131.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€131.00

About This Item

Empirical Formula (Hill Notation):
C9H6O6
CAS Number:
Molecular Weight:
210.14
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.22

€131.00


Check Cart for Availability

Request a Bulk Order

functional group

aldehyde

storage temp.

2-8°C

SMILES string

Oc1c(c(c(c(c1C=O)O)C=O)O)C=O

InChI key

KAPNIDMXEKQLMQ-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service