Skip to Content
Merck
All Photos(2)

Key Documents

392480

Sigma-Aldrich

Norepinephrine L-bitartrate hydrate

99%

Synonym(s):

α-(Aminomethyl)-3,4-dihydroxybenzyl alcohol L-bitartrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€128.00

€128.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€128.00

About This Item

Linear Formula:
(HO)2C6H3CH(CH2NH2)OH·C4H6O6 · xH2O
CAS Number:
Molecular Weight:
319.26 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€128.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

powder

optical activity

[α]27/D +12°, c = 1 in H2O

functional group

amine
carboxylic acid

SMILES string

O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.NC[C@@H](O)c1ccc(O)c(O)c1

InChI

1S/C8H11NO3.C4H6O6.H2O/c9-4-8(12)5-1-2-6(10)7(11)3-5;5-1(3(7)8)2(6)4(9)10;/h1-3,8,10-12H,4,9H2;1-2,5-6H,(H,7,8)(H,9,10);1H2/t8-;1-,2-;/m11./s1

InChI key

LNBCGLZYLJMGKP-PYYOGZMQSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service