Skip to Content
Merck
All Photos(2)

Key Documents

489350

Sigma-Aldrich

L-(–)-Norepinephrine-(+)-bitartrate

α,β-Adrenergic agonist.

Synonym(s):

L-(−)-Norepinephrine-(+)-bitartrate salt monohydrate, R(–)-4-(2-Amino-1-hydroxyethyl)-1,2-benzenediol-(+)-tartrate, Noradrenaline

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€65.40

€65.40


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€65.40

About This Item

Empirical Formula (Hill Notation):
C8H11NO3 · C4H6O6 · H2O
CAS Number:
Molecular Weight:
337.28
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€65.40


Check Cart for Availability

Request a Bulk Order

Quality Level

description

Merck USA index - 14, 6695

Assay

≥97% (LC/MS)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

NC[C@H](O)c1cc(c(cc1)O)O.O[C@H]([C@@H](O)C(=O)O)C(=O)O.O

InChI

1S/C8H11NO3.C4H6O6.H2O/c9-4-8(12)5-1-2-6(10)7(11)3-5;5-1(3(7)8)2(6)4(9)10;/h1-3,8,10-12H,4,9H2;1-2,5-6H,(H,7,8)(H,9,10);1H2/t8-;1-,2-;/m01./s1

InChI key

LNBCGLZYLJMGKP-LUDZCAPTSA-N

General description

α,β-Adrenergic agonist. A vasoconstrictive agent that causes down-regulation of angiotensin II receptors. Oxidation sensitive.

Biochem/physiol Actions

Cell permeable: no
Primary Target
α,β-Adrenergic agonist
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Highly Toxic & Carcinogenic / Teratogenic (I)

Preparation Note

Addition of sodium bisulfite (at a final concentration of 0.1%) is necessary to prevent oxidation.

Reconstitution

Following reconstituiton, refrigerate (4°C). Stock solutions are stable for up to 5 days at 4°C.

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Yang, H., et al. 1996. Hypertension 27, 1277.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Yang et al.
Hypertension (Dallas, Tex. : 1979), 27(6), 1277-1283 (1996-06-01)
Norepinephrine causes downregulation of angiotensin II (Ang II) receptors in Wistar-Kyoto rat (WKY) brain neuronal cultures. The aim of this study was to compare the cross talk between Ang II and alpha1-adrenergic receptors in these neuronal cultures. Norepinephrine causes a
Inbal Rachmin et al.
Experimental dermatology, 30(4), 578-587 (2021-02-19)
Hair greying depends on the altered presence and functionality of hair follicle melanocytes. Melanocyte stem cells (MelSCs) reside in the bulge of hair follicles and give rise to migrating and differentiating progeny during the anagen phase. Ageing, genotoxic stress, redox

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service