Skip to Content
Merck
All Photos(1)

Key Documents

392162

Sigma-Aldrich

Phenyl methacrylate

90%

Sign Into View Organizational & Contract Pricing

Select a Size

15 ML
€227.00

€227.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
15 ML
€227.00

About This Item

Linear Formula:
H2C=C(CH3)CO2C6H5
CAS Number:
Molecular Weight:
162.19
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

€227.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

90%

impurities

3% acryloyl chloride
3% phenol

refractive index

n20/D 1.512 (lit.)

bp

95-100 °C/16 mmHg (lit.)

density

1.052 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)Oc1ccccc1

InChI

1S/C10H10O2/c1-8(2)10(11)12-9-6-4-3-5-7-9/h3-7H,1H2,2H3

InChI key

QIWKUEJZZCOPFV-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olesya Daikos et al.
Physical chemistry chemical physics : PCCP, 18(47), 32369-32377 (2016-11-18)
Pentabrominated and fluorinated aromatic (meth)acrylates as well as their non-halogenated counterparts have been studied with the aim to avoid conventional photoinitiators and to overcome some negative consequences related to their use. Therefore, RTIR spectroscopy, laser flash photolysis and GC/MS were

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service