Skip to Content
Merck
All Photos(1)

Key Documents

391794

Sigma-Aldrich

1,1′-(4,6-Dihydroxy-1,3-phenylene)bisethanone

99%

Synonym(s):

4,6-Diacetylresorcinol

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€100.00

€100.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€100.00

About This Item

Linear Formula:
(HO)2C6H2(COCH3)2
CAS Number:
Molecular Weight:
194.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€100.00


Check Cart for Availability

Request a Bulk Order

Assay

99%

mp

178-180 °C (lit.)

functional group

ketone

SMILES string

CC(=O)c1cc(C(C)=O)c(O)cc1O

InChI

1S/C10H10O4/c1-5(11)7-3-8(6(2)12)10(14)4-9(7)13/h3-4,13-14H,1-2H3

InChI key

GEYCQLIOGQPPFM-UHFFFAOYSA-N

General description

1,1′-(4,6-Dihydroxy-1,3-phenylene)bisethanone (4,6-diacetylresorcinol, DAR) is a bifunctional carbonyl compound.[1] Its synthesis by acetylating resorcinol in the presence of zinc chloride has been reported.[2] The crystal structure of DAR has been studied.[3]

Application

1,1′-(4,6-Dihydroxy-1,3-phenylene)bisethanone (4,6-diacetylresorcinol, DAR) may be used in the synthesis of the following:
  • Schiff base ligands[4][1][5][6]
  • hexadentate chalcogenated bisimine ligands[7]
  • 1,5-benzodiazepines[8]
  • ketimine of chitosan[9]
  • mannich bases[10]
  • hydrazone ligands[11]
  • thiosemicarbazone, semicarbazone and thiocarbohydrazone ligands[12]
  • binuclear cobalt(II) and copper(II) complexes[13]
  • europium (III) complexes[14]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammed Sardaryar Khan et al.
Acta poloniae pharmaceutica, 67(3), 261-266 (2010-06-09)
In the present study, a series of Mannich bases was synthesized by condensing 4,6-diacetylresorcinol with formaldehyde and some selected secondary amines following the Mannich reaction conditions. Findings revealed that Mannich reaction did not take place at the acetyl function but
Magdy Shebl et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 113, 356-366 (2013-06-08)
Reactions of 4,6-diacetylresorcinol with different cobalt(II) and copper(II) salts viz., OAc(-), Cl(-), NO3(-) and SO4(2-), yielded a new series of binuclear metal complexes. Reactions of the ligand with these metal ions in the presence of a secondary ligand (L') [O,O-donor;
Design, Synthesis and Characterization of Bimetallic Palladium Complexes for Terminal Olefin Epoxidation
Netalkar SP, et al.
Catalysis Letters, 144(9), 1573-1583 (2014)
Spectral characterization and biological evaluation of Schiff bases and their mixed ligand metal complexes derived from 4, 6-diacetylresorcinol.
Pandya JH, et al.
Journal of Saudi Chemical Society, 18(3), 190-199 (2011)
Magdy Shebl
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 70(4), 850-859 (2007-11-13)
A tetradentate N2O2 donor Schiff base ligand, H2L, was synthesized by the condensation of 4,6-diacetylresorcinol with benzylamine. The structure of the ligand was elucidated by elemental analyses, IR, 1H NMR, electronic and mass spectra. Reaction of the Schiff base ligand

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service