Skip to Content
Merck
All Photos(2)

Key Documents

379743

Sigma-Aldrich

Ammonium oxalate monohydrate

≥99.99% trace metals basis

Synonym(s):

Oxalic acid diammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€196.00

€196.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
€196.00

About This Item

Linear Formula:
(NH4)2C2O4 · H2O
CAS Number:
Molecular Weight:
142.11
Beilstein:
3759830
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

€196.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.99% trace metals basis

form

powder

impurities

≤100.0 ppm Trace Metal Analysis

mp

133 °C (dec.) (lit.)

SMILES string

OC(C(O)=O)=O.N.O.N

InChI

1S/C2H2O4.2H3N.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);2*1H3;1H2

InChI key

MSMNVXKYCPHLLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Solubility of the Sodium and Ammonium Salts of Oxalic Acid in Water with Ammonium Sulfate.: This study investigates the solubility properties of ammonium oxalate monohydrate in combination with ammonium sulfate, providing insights useful for various chemical synthesis applications (Buttke et al., 2016).
  • Isolation and Identification of the Coal-Solubilizing Agent Produced by Trametes versicolor.: This research identifies ammonium oxalate monohydrate as a key agent in coal solubilization processes, highlighting its potential in industrial and environmental chemistry (Cohen et al., 1990).

Other Notes

Hydration may vary slightly

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuancun Qiao et al.
The journal of physical chemistry letters, 6(14), 2755-2760 (2015-08-13)
Negative linear compressibility (NLC) is a relatively uncommon phenomenon and rarely studied in organic systems. Here we provide the direct evidence of the persistent NLC in organic mineral ammonium oxalate monohydrate under high pressure using synchrotron X-ray powder diffraction, Raman
Electrochemiluminescence as a tool for microscopy at the nanoscale.
Lee A, et al.
Proceedings of SPIE, 13, 13-20 (2004)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service