Skip to Content
Merck
All Photos(1)

Key Documents

376310

Sigma-Aldrich

trans-3-(Trimethylsilyl)allyl alcohol

96%

Synonym(s):

trans-(3-Hydroxy-2-propen-1-yl)trimethylsilane, trans-3-(Trimethylsilyl)-2-propen-1-ol

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€187.00

€187.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€187.00

About This Item

Linear Formula:
(CH3)3SiCH=CHCH2OH
CAS Number:
Molecular Weight:
130.26
Beilstein:
1902497
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€187.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

form

liquid

impurities

<3.5% 3-trimethylsilyl-1-propanol

refractive index

n20/D 1.444 (lit.)

bp

166-168 °C (lit.)

density

0.848 g/mL at 25 °C (lit.)

functional group

aldehyde

storage temp.

−20°C

SMILES string

C[Si](C)(C)C\C=C\O

InChI

1S/C6H14OSi/c1-8(2,3)6-4-5-7/h4-5,7H,6H2,1-3H3/b5-4+

InChI key

JKKTXDCTGMUFBS-SNAWJCMRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-3-(Trimethylsilyl)allyl alcohol is a 3-silyl allyl alcohol.

Application

trans-3-(Trimethylsilyl)allyl alcohol is suitable for use in the preparation of 1-iodo-3-trimethylsilyl-2-propene, via mesylation followed by iodide displacement.[1]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

136.4 °F - closed cup

Flash Point(C)

58 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minh Pham et al.
The Journal of organic chemistry, 73(2), 741-744 (2007-12-22)
In the presence of indium metal, 3-iodo-2-[(trimethylsilyl)methyl]propene (1) reacts with sequentially added aldehydes to provide cis-2,6-disubstituted tetrahydropyrans in good yields. Evidence suggests that InI, formed upon aldehyde (R1CHO) allylation in aqueous media, acts as a promoter for the silyl-Prins reaction

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service