Skip to Content
Merck
All Photos(2)

Key Documents

370460

Sigma-Aldrich

Diethoxyacetonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2CHCN
CAS Number:
Molecular Weight:
129.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

form

liquid

refractive index

n20/D 1.4 (lit.)

bp

167.7 °C/773 mmHg (lit.)

mp

−19-−18 °C (lit.)

density

0.929 g/mL at 25 °C (lit.)

functional group

ether
nitrile

SMILES string

CCOC(OCC)C#N

InChI

1S/C6H11NO2/c1-3-8-6(5-7)9-4-2/h6H,3-4H2,1-2H3

InChI key

UDELMRIGXNCYLU-UHFFFAOYSA-N

Related Categories

General description

Diethoxyacetonitrile is a 2,2-dialkoxyalkanenitrile derivative. Reaction of hydrocyanic acid with various ortho esters of aliphatic and aromatic acids in the presence of acidic catalyst has been reported to afford diethoxyacetonitrile.[1] Houben-Hoesch reaction of diethoxyacetonitrile with 1,2,4-trimethoxybenzene affords tris(2,4,5-trimethoxyphenyl)methane.[2]

Application

Diethoxyacetonitrile may be used in the preparation of methyl 5-diethoxymethylimidazole-4-carboxylate, via anionic cycloaddition reaction with methyl isocyanoacetate.[3]

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An efficient synthesis of 5-diethoxymethylimidazole-4-carboxylate, a potential precursor for various imidazole derivatives.
Murakami T, et al.
Tetrahedron Letters, 23(45), 4729-4732 (1982)
2, 2-Dialkoxyalkanenitriles.
Erickson JG.
Journal of the American Chemical Society, 73(3), 1338-1339 (1951)
An anomalous Houben-Hoesch reaction and some applications in arylation reactions.
Sanchez-Viesca F, et al.
Organic Prep. and Proc. Int., 36(2), 135-140 (2004)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service