Skip to Content
Merck
All Photos(2)

Key Documents

365297

Sigma-Aldrich

Lithium metaborate dihydrate

Synonym(s):

Lithium borate (LiBO2 ) dihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€125.00

€125.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 G
€125.00

About This Item

Linear Formula:
LiBO2 · 2H2O
CAS Number:
Molecular Weight:
85.78
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid

€125.00


Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

concentration

11.7-13% B (by NaOH, titration)

SMILES string

[Li+].O.O.[O-]B=O

InChI

1S/BO2.Li.2H2O/c2-1-3;;;/h;;2*1H2/q-1;+1;;

InChI key

MAXQFCAELLTPNS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service