Skip to Content
Merck
All Photos(1)

Key Documents

367559

Sigma-Aldrich

(S)-(+)-1-Aminoethylphosphonic acid

99%

Synonym(s):

D-(+)-1-Aminoethylphosphonic acid, D-Ala(P)

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€265.00

€265.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€265.00

About This Item

Linear Formula:
CH3CH(NH2)P(O)(OH)2
CAS Number:
Molecular Weight:
125.06
Beilstein:
4291033
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

€265.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

solid

optical activity

[α]20/D +4.8°, c = 5 in H2O

mp

290 °C (dec.) (lit.)

functional group

amine

SMILES string

C[C@@H](N)P(O)(O)=O

InChI

1S/C2H8NO3P/c1-2(3)7(4,5)6/h2H,3H2,1H3,(H2,4,5,6)/t2-/m0/s1

InChI key

UIQSKEDQPSEGAU-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Mironenko et al.
Biokhimiia (Moscow, Russia), 55(6), 1124-1131 (1990-06-01)
Penicillin acylase from E. coli (EC 3.5.1.11) was found to hydrolyze N-phenylacetylated 1-aminoethylphosphonic acid and its esters. The enzyme preferentially converts the R-form of the substrates: the ratios of the bimolecular rate constants of penicillin acylasecatalyzed hydrolysis of R- and
O Ploux et al.
European journal of biochemistry, 259(1-2), 63-70 (1999-01-23)
8-Amino-7-oxopelargonate synthase catalyzes the first committed step of biotin biosynthesis in micro-organisms and plants. Because inhibitors of this pathway might lead to antibacterials or herbicides, we have undertaken an inhibition study on 8-amino-7-oxopelargonate synthase using six different compounds. d-Alanine, the
V Copié et al.
Biochemistry, 27(14), 4966-4970 (1988-07-12)
Inhibition of alanine racemase from the Gram-positive bacterium Bacillus stearothermophilus by (1-aminoethyl) phosphonic acid (Ala-P) proceeds via a two-step reaction pathway in which reactivation occurs very slowly. In order to determine the mechanism of inhibition, we have recorded low-temperature, solid-state
F R Atherton et al.
Journal of medicinal chemistry, 29(1), 29-40 (1986-01-01)
Phosphonodipeptides and phosphonooligopeptides based on L- and D-(1-aminoethyl)phosphonic acids L-Ala(P) and D-Ala(P) and (aminomethyl)phosphonic acid Gly(P) at the acid terminus have been synthesized and investigated as antibacterial agents, which owe their activity to the inhibition of bacterial cell-wall biosynthesis. A

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service