Skip to Content
Merck
All Photos(1)

Key Documents

366358

Sigma-Aldrich

(R)-(−)-5-(Hydroxymethyl)-2-pyrrolidinone

99%

Synonym(s):

D-Pyroglutaminol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
3587974
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
99%

Assay

99%

form

solid

optical activity

[α]20/D −31°, c = 5 in ethanol

bp

147-149 °C/0.06 mmHg (lit.)

mp

83-85 °C (lit.)

functional group

hydroxyl

SMILES string

OC[C@H]1CCC(=O)N1

InChI

1S/C5H9NO2/c7-3-4-1-2-5(8)6-4/h4,7H,1-3H2,(H,6,8)/t4-/m1/s1

InChI key

HOBJEFOCIRXQKH-SCSAIBSYSA-N

Application

Serves as a building block in the synthesis of diphthamide.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of the American Chemical Society, 114, 1495-1495 (1992)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service