Skip to Content
Merck
All Photos(1)

Key Documents

360139

Sigma-Aldrich

5′-O-(4,4′-Dimethoxytrityl)thymidine

98%

Synonym(s):

DMT-T

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€286.00

€286.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€286.00

About This Item

Empirical Formula (Hill Notation):
C31H32N2O7
CAS Number:
Molecular Weight:
544.59
Beilstein:
599297
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€286.00


Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

114-116 °C (subl.) (lit.)

functional group

ether
hydroxyl
phenyl

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H](C[C@@H]2O)N3C=C(C)C(=O)NC3=O)(c4ccccc4)c5ccc(OC)cc5

InChI

1S/C31H32N2O7/c1-20-18-33(30(36)32-29(20)35)28-17-26(34)27(40-28)19-39-31(21-7-5-4-6-8-21,22-9-13-24(37-2)14-10-22)23-11-15-25(38-3)16-12-23/h4-16,18,26-28,34H,17,19H2,1-3H3,(H,32,35,36)/t26-,27+,28+/m0/s1

InChI key

UBTJZUKVKGZHAD-UPRLRBBYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5′-O-(4,4′-Dimethoxytrityl)thymidine is a pyrimidine.

Application

5′-O-(4,4′-Dimethoxytrityl)thymidine may be used in the stereoselective synthesis of 3′-deoxy-3′-threo-hydroxymethyl nucleoside, which can be subsequently incorporated into oligodeoxynucleotides.[1] It may be used as research tool for antiviral and anticancer studies.[2]

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oligodeoxynucleotide analogues containing 3'-deoxy-3'-C- threo-hydroxymethylthymidine: Synthesis, hybridization properties and enzymatic stability.
Svendsen ML, et al.
Tetrahedron, 49(48), 11341-11352 (1993)
Tamara I Prykota et al.
Nucleosides, nucleotides & nucleic acids, 30(7-8), 544-551 (2011-09-06)
A new labeling technique attaching a fluorescent pteridine derivative (3, 5) via a linker onto the 3'-OH group of 5'-O-dimethoxytritylthymidine (7) was developed to lead to the conjugates 8 and 11. After detritylation to give 9 and 12, the final
K Miyoshi et al.
Nucleic acids research, 8(22), 5473-5489 (1980-11-25)
Synthesis of two oligothymidylic acids, tridecamer and nonadecamer, is described by a rapid and simple solid-phase method on two kinds of polyacrylamide supports derivatized from commercially available Enzacryl Gel K-2. The syntheses were performed by the phosphotriester method using di-
A Elmblad et al.
Nucleic acids research, 10(10), 3291-3301 (1982-05-25)
A method for the synthesis of mixed dimers, trimers and oligonucleotides on a solid support using monomeric protected nucleoside phosphochloridites (1a-d) has been developed and the different nucleoside reagents, and the results show that yields of different oligomers in a
A Guzaev et al.
Bioorganic & medicinal chemistry letters, 8(9), 1123-1126 (1999-01-01)
A novel solid-phase synthesis of 5'-radiolabeled oligonucleotides is described. The labeling reaction is carried out by the phosphoramidite method with the aid of [4,6-di-14C]-5'-dimethoxytritylthymidine building block 1. The feasibility of the method is demonstrated by preparation of 3'-phosphorylated dodecathymidylate phosphorothioate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service