Skip to Content
Merck
All Photos(2)

Key Documents

348554

Sigma-Aldrich

3,3′-Iminobis(N,N-dimethylpropylamine)

97%

Synonym(s):

(3-[[3-(Dimethylamino)propyl]amino]propyl)dimethylamine, 1,1,9,9-Tetramethyl-1,5,9-triazanonane, 2,10-Dimethyl-2,6,10-triazaundecane, Bis[3-(dimethylamino)propyl]amine, N,N,N′,N′-Tetramethyldipropylenetriamine, N,N,N′,N′-Tetramethyliminobispropylamine, N,N-Bis[3-(dimethylamino)propyl]amine, N-(3-Dimethylaminopropyl)-N′,N′-dimethylpropane-1,3-diamine, Tetramethyldipropylenetriamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HN[(CH2)3N(CH3)2]2
CAS Number:
Molecular Weight:
187.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
97%

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.449 (lit.)

bp

128-131 °C/20 mmHg (lit.)

mp

−78 °C (lit.)

density

0.841 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CN(C)CCCNCCCN(C)C

InChI

1S/C10H25N3/c1-12(2)9-5-7-11-8-6-10-13(3)4/h11H,5-10H2,1-4H3

InChI key

BXYVQNNEFZOBOZ-UHFFFAOYSA-N

Application

3,3′-Iminobis(N,N-dimethylpropylamine) may be used:
  • in the synthesis of dimeric quaternary alkylammonium conjugates of sterols
  • as nitrogen containing tridentate lignand in the preparation of [3,3′-iminobis(N,N-dimethylpropylamine)](4′-methoxyflavonolato)zinc(II) perchlorate complex
  • in the preparation of 2-[[[N,N-bis[3-(N,N-dimethylamino)propyl]amino]carbonyl]-1-methyl-4-nitropyrrole and 3-(9-anthracenyl)- N,N-bis[3-(N,N-dimethylamino)propyl]-5-methyl-4-isoxazole carboxamide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaochun Han et al.
Bioorganic & medicinal chemistry, 17(4), 1671-1680 (2009-01-27)
The synthesis and in vitro anti-tumor 60 cell lines screen of a novel series of anthracenyl isoxazole amides (AIMs) (While not a strict acronym, the designation AIM is in honor of the memory of Professor Albert I. Meyers.) (22-33) are
Bogumił Brycki et al.
Molecules (Basel, Switzerland), 19(7), 9419-9434 (2014-07-06)
New dimeric quaternary alkylammonium conjugates of sterols were obtained by two step reactions of ergosterol, cholesterol and cholestanol with bromoacetic acid bromide, followed by bimolecular nucleophilic substitution with N,N,N',N'-tetramethyl-1,3-propanediamine, N,N,N',N'',N''-pentamethyldiethylenetriamine and 3,3'-iminobis- (N,N-dimethylpropylamine). The product structures were confirmed by spectral
[3, 3'-Iminobis (N, N-dimethylpropylamine)](4'-methoxyflavonolato) zinc (II) perchlorate,[Zn (4'-MeOfla)(idpaH)] ClO4.
Balogh-Hergovich E, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 55(4), 557-558 (1999)
Łukasz Łątka et al.
Materials (Basel, Switzerland), 14(4) (2021-02-11)
Polyamide-based nanocomposites containing graphene platelets decorated with poly(acrylamide) brushes were prepared and characterized. The brushes were grafted from the surface of graphene oxide (GO), a thermally conductive additive, using atom transfer radical polymerization, which led to the formation of the
[Toxicity of several aliphatic amines].
G I Sidorin et al.
Gigiena truda i professional'nye zabolevaniia, (11)(11), 50-53 (1984-11-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service